Learn More
Creative Biomart Eftrenonacog alfa, Recombinant Factor IX
Supplier:  Creative Biomart THP0066
Encompass_Preferred
The product is comprised of a single molecule of human factor IX (FIX) covalently linked to the constant region (Fc) domain of human IgG1 via recombinant DNA technology in a human embryonic kidney cell line (HEK293H).
Catalog No. 50-208-4117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.