Learn More
Abcam Recombinant Hepatitis C Virus NS4A protein, 10UG
Supplier: Abcam AB8435010UG
Recombinant Hepatitis C Virus NS4A protein, 10UG
Protein/ Peptide/ Enzyme Synonyms: Genome polyprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.