Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Recombinant Hepatitis C Virus NS3 Protein, >95% Purity, 100 ug

Catalog No. 502710692
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0692 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0692 Supplier Abcam Supplier No. AB49025100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Hepatitis C virus NS3 protein fragment with >95% purity for use in ELISA, SDS-PAGE, WB, and flow cytometry.

  • Protein Type: Recombinant Hepatitis C Virus NS3 protein (isolate BK)
  • Expression System: Escherichia coli
  • Amino Acid Range: 1450 to 1643 aa
  • Purity: >95%
  • Applications: ELISA, Flow Cyt, SDS-PAGE, WB, FuncS
  • Formulation: Liquid. pH 7.2 to 7.6. Constituents include 50% glycerol, 48% urea, 0.316% Tris HCl, and 0.078% 2-mercaptoethanol
  • Concentration: 1 mg/mL

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Hepatitis C Virus NS3
Shipping Condition Blue Ice
Buffer 50% Glycerol, 48% urea, 0.316% Tris-HCl, 0.078% 2-mercaptoethanol, pH7.2 to pH7.6
Label or Dye Unconjugated
Concentration 1 mg/mL
Content And Storage Store at -20°C.
Form Liquid
Formulation 50% Glycerol, 48% Urea, 0.316% Tris HCl, 0.078% 2-Mercaptoethanol, pH 7.2 to 7.6
Quantity 100 μg
For Use With (Application) SDS-PAGE, ELISA, Western Blot, Flow Cytometry, FuncS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.