Learn More
Abcam Recombinant Hepatitis C Virus NS3 Protein, E. coli, 100 μg
Description
- Target Protein: Hepatitis C Virus NS3
- Expression System: Escherichia coli
- Purity: >95%
- Tested Applications: ELISA, Flow Cyt, SDS-PAGE, WB
- Formulation: Liquid, pH 7.2 to 7.6
- Constituents: 50% Glycerol, 9% Urea, 0.395% Tris HCl, 0.2% Triton X-100
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Hepatitis C Virus NS3 |
| Substrate Properties | Protein-based Substrate |
| Shipping Condition | Blue Ice |
| Label or Dye | Unconjugated |
| Buffer | 50% glycerol, 9% urea, 0.395% Tris-HCl, 0.2% Triton X-100, pH7.2 to pH7.6 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Formulation | 50% Glycerol, 9% Urea, 0.395% Tris HCl, 0.2% Triton X-100, pH 7.2 to 7.6 |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.