Learn More
Abcam Recombinant Hepatitis C Virus NS4 Protein, E. coli, 100 ug
Description
- Protein Type: Recombinant
- Source: Escherichia coli
- Target: Hepatitis C virus (isolate BK) NS4 protein (1916–1947 aa)
- Purity: >95%
- Applications: ELISA, Flow Cyt, SDS-PAGE, WB
- Formulation: Liquid, pH 7.2–7.6, containing 50% glycerol, 9% urea, 0.395% Tris HCl, and 0.2% Triton X-100
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Liquid |
| Target Molecule | Hepatitis C Virus (NS4 Protein) |
| Buffer | 50% glycerol, 9% urea, 0.395% Tris-HCl, 0.2% Triton X-100, pH7.2 to pH7.6 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Label or Dye | Unconjugated |
| Reactivity | Hepatitis virus |
| Shipping Condition | Blue Ice |
| Formulation | 50% Glycerol (glycerin, glycerine), 9% Urea, 0.395% Tris HCl, 0.2% Triton X-100, pH 7.2 - 7.6 |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.