Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Recombinant Hepatitis C Virus NS5 Protein, E. Coli, >95% Purity, 100 μg

Catalog No. 502710687
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0687 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0687 Supplier Abcam Supplier No. AB49032100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant protein expressed in E. coli with >95% purity for use in ELISA, WB, and Flow Cyt.

  • Protein Type: Recombinant Hepatitis C Virus NS5 protein (isolate EUH1480, aa 2061 to 2302)
  • Expression System: Escherichia coli
  • Purity: >95%
  • Tested Applications: ELISA, Flow Cyt, WB
  • Formulation: Liquid, pH 7.2 to 7.6; contains 50% glycerol, 9% urea, 0.395% Tris HCl, and 0.2% Triton X-100
  • Concentration: 1 mg/mL

Specifications

Target Molecule Hepatitis C Virus NS5
Shipping Condition Blue Ice
Label or Dye Unconjugated
Form Liquid
Content And Storage Store at -20°C.
Concentration 1 mg/mL
Buffer 50% glycerol, 9% urea, 0.395% Tris-HCl, 0.2% Triton X-100, pH7.2 to pH7.6
Formulation 50% Glycerol, 9% Urea, 0.395% Tris HCl, 0.2% Triton X-100, pH 7.2 - 7.6
Quantity 100 μg
For Use With (Application) ELISA, Western Blot, Flow Cyt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.