Learn More
Active Motif Recombinant Histone H1
SDP
Supplier:  Active Motif 81826
Encompass
Recombinant Histone H1 is suitable for use as substrates in the study of enzyme kinetics, inhibitor screening, and selectivity profiling. Full length Histone H1 (accession number NP_005309.1) was expressed in E. coli cells with no tag. The molecular weight of Histone H1 is 21 kDa. For more complete information, please visit the Active Motif website.
Catalog No. NC1849789
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.