Learn More
R&D Systems™ Recombinant HIV-1 Gag p24 His-tag Protein
Description
Specifications
Specifications
| Accession Number | NP_057850.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS, Urea with Trehalose. |
| Molecular Weight (g/mol) | M.W.-observed: 21-25 kDa, under reducing conditions., M.W.-theroretical: 24 kDa |
| Quantity | 100 μg |
| Source | E. coli-derived viral HIV-1 Gag p24 protein Pro149-Gly354, with a C-terminal 6-His tag and a N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Purity or Quality Grade | >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.