Learn More
Abcam Recombinant HPV16 L1 protein, 50UG
Supplier:  Abcam AB11988050UG
Encompass_Preferred
Recombinant HPV16 L1 protein is a Human papillomavirus type 16 Full Length protein, in the 1 to 505 aa range, expressed in Saccharomyces cerevisiae, with >90% purity and suitable for SDS-PAGE, EM, WB.
Protein/ Peptide/ Enzyme Synonyms: Major capsid protein L1, L1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2135437
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.