Learn More
Specifications
Specifications
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | 10 mM Tris (pH 8), 105 mM NaCl, 5 mM EDTA |
| Gene ID (Entrez) | 886354 |
| Molecular Weight (g/mol) | 65 kDa |
| Quantity | 1 x 100 μg Vials |
| Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | 60 kDa chaperonin 2, 65 kDa antigen, Antigen A, Cell wall protein A, GroEL protein 2, groEL-2, groL2, Heat shock protein 65, hsp65, MT0456, MTV037.04, Protein Cpn60-2, Rv0440 |
| Gene Symbol | groEL2 |
| Purity or Quality Grade | >90%, by SDS-PAGE |
| Protein | HSP65 (mycobacterial) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
