Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human 14-3-3 epsilon Protein
SDP

Catalog No. NBC118392 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118392 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118392 Supplier Novus Biologicals™ Supplier No. NBC118392
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids1-255 of Human 14-3-3 epsilon The Recombinant Human 14-3-3 epsilon Protein is derived from E. coli. The Recombinant Human 14-3-3 epsilon Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P62258
For Use With (Application) SDS-PAGE
Formulation 20mM Tris buffer (pH 7.5)
Gene ID (Entrez) 7531
Molecular Weight (g/mol) 29kDa
Name 14-3-3 epsilon Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 14-3-3 epsilon, 1-255aa, Human, Recombinant, E.coli (NP_006752).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.