Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human 14-3-3 tau/theta Protein
SDP

Catalog No. NBC118393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118393 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118393 Supplier Novus Biologicals™ Supplier No. NBC118393
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids1-245 of Human 14-3-3 tau/theta The Recombinant Human 14-3-3 tau/theta Protein is derived from E. coli. The Recombinant Human 14-3-3 tau/theta Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P27348
For Use With (Application) Western Blot
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 10971
Molecular Weight (g/mol) 28kDa
Name 14-3-3 tau/theta Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 14-3-3 tau/theta, 1-245aa, Human, Recombinant, E.coli (NP_006817).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.