Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human 4-1BB Ligand/TNFSF9 Protein
GSA_VA

Catalog No. 22954L025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and PEG and with Trehalose.
Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and PEG with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
22954L025 Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and PEG with BSA as a carrier protein.
22954L025CF Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and PEG and with Trehalose.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22954L025 Supplier R&D Systems™ Supplier No. 22954L025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Human (rh) 4-1BB/TNFRSF9/CD137 Fc Chimera (Catalog # 838-4B ) is Immobilized at 10 ng/mL (100 μL/well), the concentration of rh4-1BB Ligand that produces 50% optimal binding response is found to be approximately 0.5-2.5 ng/mL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accession Number P41273.1
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and PEG with BSA as a carrier protein.
Gene ID (Entrez) 8744
Molecular Weight (g/mol) M.W. (theoretical): 20.4 kDa
Quantity 25 μg
Source E. coli-derived human 4-1 BB Ligand/TNFSF9 protein Arg71-Glu254, with an N-terminal Met and a 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, 2°C to 8°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFSF9
Product Type 4-1BB Ligand/TNFSF9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein 4-1 BB Ligand/TNFSF9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.