Learn More
R&D Systems™ Recombinant Human 5'-Nucleotidase/CD73 Fc Avi-tag, CF
Description
Specifications
Specifications
| Accession Number | AAH65937.1 |
| For Use With (Application) | Enzyme Activity, Binding Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2 and Glycerol. |
| Gene ID (Entrez) | 4907 |
| Molecular Weight (g/mol) | M.W. (Observed): 88-98 kDa, under reducing conditions, M.W. (Theroretical): 86 kDa |
| Quantity | 25 μg |
| Source | Human embryonic kidney cell, HEK293-derived human 5'-Nucleotidase/CD73 protein Human 5'-Nucleotidase/CD73 (Trp27-Lys547) (N-terminus) Accession # AAH65937.1 GGIEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.