Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human 5'-Nucleotidase/CD73 Protein
SDP

Catalog No. 5795EN010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5795EN010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5795EN010 Supplier R&D Systems™ Supplier No. 5795EN010
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to hydrolyze the 5’-phosphate group from the substrate adenosine-5’-monophosphate (AMP). The orthophosphate product is measured by a Malachite Green Phosphate Detection Kit (Catalog # DY996 ). The specific activity is >15,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number AAH65937
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2 and Glycerol.
Gene ID (Entrez) 4907
Molecular Weight (g/mol) M.W. (Observed): 61-62 kDa, reducing conditions; M.W. (theoretical): 59 kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human 5 '-Nucleotidase/CD73 protein Trp27-Lys547, with a C-terminal 6-His tag, Trp27-Lys547
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Structural Form Dimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol NT5E
Product Type 5'-Nucleotidase/CD73
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein 5 '-Nucleotidase/CD73
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.