Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human 5T4 Fc Chimera Protein
SDP

Catalog No. 4975TG100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4975TG100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4975TG100 Supplier R&D Systems™ Supplier No. 4975TG100
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human 5T4 Fc Chimera Protein is derived from CHO. The Recombinant Human 5T4 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q13641
For Use With (Application) Bioactivity
Gene ID (Entrez) 7162
Molecular Weight (g/mol) 62 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line,CHO-derived human 5T4 protein, (N-terminus) Human 5T4 (Ser31-Ser355) Accession &Num; Q13641 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Specific Reactivity 5T4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 1 mg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.