Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human A33/GPA33 His (C-Term) Protein
SDP

Catalog No. NB171823 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171823 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171823 Supplier Novus Biologicals™ Supplier No. NBP318201
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from a 0.22 um filtered solution of PBS, pH 7.4
Gene ID (Entrez) 10223
Quantity 100 μg
Storage Requirements Store at -20°C.
Endotoxin Concentration <0.1 EU/μg of the protein by LAL method
Gene Alias A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33
Gene Symbol GPA33
Reconstitution Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water
Research Category Cancer
Purity or Quality Grade >95%, by SDS-PAGE
Protein A33/GPA33
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.