Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ACE-2 Protein

Catalog No. 933ZN010
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
933ZN010 10 μg
933ZN100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 933ZN010 Supplier R&D Systems™ Supplier No. 933ZN010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its binding ability in a functional ELISA. When Recombinant SARS-CoV-2 Spike RBD Fc Chimera (Catalog # 10499-CV ) is immobilized at 0.2 μg/mL (100 μL/well), Recombinant Human ACE-2 (Catalog # 933-ZN) binds with an ED50 of 1.00-12.0 ng/mL. Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH (Catalog # ES007 ). The specific activity is >800 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q9BYF1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, ZnCl2 and Glycerol.
Gene ID (Entrez) 59272
Molecular Weight (g/mol) M.W. (Observed): 101-111 kDa, reducing conditions; M.W. (theoretical): 85 kDa
Name ACE-2
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human ACE-2 protein Gln18-Ser740, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Recombinant Human ACE-2 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol ACE2
Conjugate Unconjugated
Recombinant Recombinant
Activity Measured by its binding ability in a functional ELISA with Recombinant Viral SARS-CoV-2 S Protein RBD Fc Chimera (Catalog # 10499-CV). Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH (Catalog # ES007 ). The specific activity is >800 pmol/min/μg, as measured under the described conditions.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein ACE-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.