Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Active Heparanase/HPSE Protein

Catalog No. 7570GH005
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7570GH005 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 7570GH005 Supplier R&D Systems™ Supplier No. 7570GH005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to release biotinylated heparan sulfate from Recombinant Human Syndecan-4 (Catalog # 2918-SD ). 20 ng of Recombinant Human Active Heparanase/HPSE digestion will result in >50% of OD reduction compared with the Negative Control, as measured under the described conditions.

Specifications

Accession Number AF144325
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and E64.
Gene ID (Entrez) 10855
Molecular Weight (g/mol) M.W. (Observed): 50-60 kDa & 7-9 kDa, reducing conditions; M.W. (theoretical): 43 kDa & 9 kDa
Name Active Heparanase/HPSE
Quantity 5 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Heparanase/HPSE protein Gln36-Ile543 with N-terminal 6-His tag The linker peptide from Ser110-Gln157 is removed by protease treatment
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol HPSE
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Heparanase/HPSE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.