Learn More
Description
Specifications
Specifications
| Accession Number | Q02750.2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 5604 |
| Molecular Weight (g/mol) | M.W.-observed: 62-68 kDa, under reducing conditions, M.W.-theroretical: 69 kDa |
| Quantity | 20 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human MEK1 protein Met1-Val393 with an N-terminal GST tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAP2K1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
