Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Active p70 S6 Kinase Protein

Catalog No. 896KS010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
896KS010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 896KS010 Supplier R&D Systems™ Supplier No. 896KS010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The activity of p70S6K was determined to be 73 nmol/min/mg using a synthetic peptide substrate.

Specifications

For Use With (Application) Bioactivity
Formulation Supplied in 50mM sodium phosphate (pH 7.0), 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, and 25% glycerol.
Molecular Weight (g/mol) Observed MW: 76kDa
Quantity 10 μg
Source Spodoptera frugiperda,Sf 9 (baculovirus)-derived human p70 S6 Kinase protein
Storage Requirements This product is stable at ≤ -70° C for up to one year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles.
Conjugate Unconjugated
Content And Storage This product is stable at <= -70°C for up to one year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles.
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.