Learn More
R&D Systems™ Recombinant Human Active PDK-1 Protein
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Supplied in 50mM sodium phosphate (pH 7.0), 300mM NaCl, 0.25mM DTT, 150mM imidazole, 0.1mM PMSF, and 25% glycerol. |
| Gene ID (Entrez) | 5163 |
| Molecular Weight (g/mol) | Observed MW: 67kDa |
| Quantity | 10 μg |
| Source | Spodoptera frugiperda,Sf 9 (baculovirus)-derived human PDK-1 protein |
| Storage Requirements | This product is stable at ≤ -70° C for up to one year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles. |
| Gene Alias | EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PDPK1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial |
| Conjugate | Unconjugated |
| Content And Storage | This product is stable at <= -70°C for up to one year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.