Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Active Raf-1 (Y340E Y341E, 306-end)

Catalog No. 4540KS010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4540KS010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4540KS010 Supplier R&D Systems™ Supplier No. 4540KS010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The activity of Raf-1 is typically 5100-6900 nmol/min/mg using a myelin basic protein (MBP) substrate (see Activity Assay Protocol).

Specifications

Accession Number NM_002880
For Use With (Application) Bioactivity
Formulation Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Gene ID (Entrez) 5894
Molecular Weight (g/mol) M.W. (theoretical): 63 kDa
Quantity 10 μg
Source Spodoptera frugiperda, Sf 9 (baculovirus)-derived human Raf-1 protein aa 306-648 Accession # NM_002880
Storage Requirements This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.)
Gene Symbol RAF1
Conjugate Unconjugated
Purity or Quality Grade >70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein Raf-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.