Learn More
R&D Systems™ Recombinant Human Active SYK Protein
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Supplied in 50mM Tris-HCl (pH 7.5), 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol. |
| Molecular Weight (g/mol) | Observed MW: 100kDa |
| Quantity | 10 μg |
| Source | Spodoptera frugiperda,Sf 9 (baculovirus)-derived human SYK protein Accession &Num; NM_003177 |
| Storage Requirements | This product is stable at ≤ -70° C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.