Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Recombinant human activin A protein
Shop All Sartorius Products
Click to view available options
Quantity:
0.025 mg
0.1 mg
0.5 mg
Description
- Research Use Only

Specifications
Specifications
For Use With (Application) | Organoid Expansion |
Quantity | 0.1 mg |
Product Type | Recombinant Proteins |
Conjugate | Unconjugated |
Species | Human |
Recombinant | Recombinant |
Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction