Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Activin AC Heterodimer Protein

Catalog No. 4879AC010CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
4879AC010CF Lyophilized from a 0.2μm filtered solution in HCl.
4879AC010 Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 4879AC010CF Supplier R&D Systems™ Supplier No. 4879AC010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human Activin AC Heterodimer Protein is derived from CHO. The Recombinant Human Activin AC Heterodimer Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HCl.
Molecular Weight (g/mol) 12.9 kDa (betaA subunit), 12.5 kDa (betaC subunit)
Name Activin AC
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Activin AC protein, (N-terminus) Human Activin beta A (Gly311-Ser426) Accession &Num; NP_002183 Human Activin beta C (Gly237-Ser352) Accession &Num; NP_005529 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.