Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Activin B Protein

Catalog No. 659AB005
Change view
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
659AB005 5 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
659AB005CF 5 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose.
659AB025 25 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
659AB025CF 25 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 659AB005 Supplier R&D Systems™ Supplier No. 659AB005
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Activin B Protein is derived from CHO. The Recombinant Human Activin B Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 3625
Molecular Weight (g/mol) 12.8 kDa (monomer)
Name Activin B
Quantity 5 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Activin B protein Gly293-Ala407
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.