Learn More
R&D Systems™ Recombinant Human Activin B Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose. |
| Gene ID (Entrez) | 3625 |
| Molecular Weight (g/mol) | 12.8 kDa (monomer) |
| Name | Activin B |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Activin B protein Gly293-Ala407 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.