Learn More
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | Q13444 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in MES and NaCl |
| Gene ID (Entrez) | 8751 |
| Molecular Weight (g/mol) | 53 kDa, reducing conditions |
| Name | ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC15, MDC-15, Metargidin |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human ADAM15 protein Asp207-Ser693 with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Biological Activity | Supports adhesion of Jurkat human acute T cell leukemia cells. The ED50 0.5 to 3μg/mL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
