Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ADAMTS-L2 His-tag Protein

Catalog No. 10950AD020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10950AD020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10950AD020 Supplier R&D Systems™ Supplier No. 10950AD020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human ADAMTSL2 His-tag (Catalog# 10950-AD) immobilized at 2.00 μg/mL, 100 μL/well, the concentration of Recombinant Human Lysyl Oxidase Homolog 2 Protein (Catalog # 2639-AO ) that produces 50% of the optimal binding response is approximately 0.60-3.00 μg/mL.

Specifications

Accession Number Q86TH1.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HEPES and NaCl with Trehalose.
Gene ID (Entrez) 9719
Molecular Weight (g/mol) M.W.-observed: 130-144 kDa, under reducing conditions, M.W.-theroretical: 103 kDa
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human ADAMTSL2 protein Asp23-Ser951 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias ADAMTSL2 ADAMTS-like 2, GPHYSD1
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ADAMTSL2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.