Learn More
R&D Systems™ Recombinant Human ADAMTS4 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate, CaCl 2 and NaCl. |
| Gene ID (Entrez) | 9507 |
| Molecular Weight (g/mol) | 53 kDa |
| Name | ADAMTS4 |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human ADAMTS4 protein Phe213-Cys685,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Recombinant Human ADAMTS4 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.