Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Adiponectin/Acrp30 Protein
SDP

Catalog No. NBC118461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118461 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118461 Supplier Novus Biologicals™ Supplier No. NBC118461
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 15-244 of Human Adiponectin/Acrp30 The Recombinant Human Adiponectin/Acrp30 Protein is derived from E. coli. The Recombinant Human Adiponectin/Acrp30 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q15848
For Use With (Application) ELISA, SDS-PAGE
Formulation PBS (pH 7.4), 1mM DTT
Gene ID (Entrez) 9370
Molecular Weight (g/mol) 25.1kDa
Name Adiponectin/Acrp30 protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Adiponectin, 15-244 aa, Human, Recombinant, E.coli (NP_004788).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.