Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human AICL/CLEC-2B Fc Chimera Protein

Catalog No. 9059CL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-59C-L050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-59C-L050 Supplier R&D Systems™ Supplier No. 9059CL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human AICL/CLEC-2B Fc Chimera Protein is derived from CHO. The Recombinant Human AICL/CLEC-2B Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 9976
Molecular Weight (g/mol) Observed M.W.: 48-65kDa, reducing conditions, Theoretical M.W.: 41kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human AICL/CLEC-2B protein, (N-terminus) MD Human IgG1 (Pro100-Lys330) IEGR Human AICL/CLEC-2B (Lys26-His149) with a Met79Thr substitution Accession &Num; Q92478 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Activation-induced C-type lectin, AICLC-type lectin superfamily member 2, CLEC2B, CLEC-2B, CLECSF2, CLECSF2IFN-alpha-2b-inducing-related protein 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 2 (activation-induced), C-type lectin domain family 2 member B, C-type lectin domain family 2, member B, HP10085, IFN-alpha2b-inducing related protein 1, IFNRG1
Common Name AICL
Gene Symbol CLEC2B
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human AICL/CLEC-2B Fc Chimera is coated at 2μg/mL (100μL/well), the concentration of When Recombinant Human AICL/CLEC-2B Fc Chimera is coated at 2μg/mL (100μL/well), the concentration of Recombinant Human Galectin-1 (Catalog # 1152-GA) that produces 50% optimal binding response is typically 0.6-3μg/mL. that produces 50% optimal binding response is typically 0.6-3μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived MD, N-terminus; Human IgG1 (Pro100-Lys330); IEGR; Human AICL/CLEC-2B (Lys26-His149) with a Met79Thr substitution Accession # Q92478, C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.