Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 57016 |
| Molecular Weight (g/mol) | 37 kDa |
| Name | Aldo-keto Reductase 1B10/AKR1B10 |
| Quantity | 20 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Aldo-keto Reductase 1B10/AKR1B10 protein Ala2-Tyr316,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
