Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ALK/CD246 His-tag Protein

Catalog No. 4210CD100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4210CD100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4210CD100 Supplier R&D Systems™ Supplier No. 4210CD100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. In a Human ALK/CD246 Antibody (Catalog # AF4210) coated plate, when Recombinant Human ALK/CD246 is present at 0.5 μg/mL, Recombinant Human Pleiotrophin/PTN (Catalog # 252-PL ) binds with an ED50 of 20-100 ng/mL.

Specifications

Accession Number Q9UM73.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 238
Molecular Weight (g/mol) M.W.-observed: 130-150 kDa, under reducing conditions, M.W.-theroretical: 111 kDa
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived human ALK/CD246 protein Val19-Ser1038, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, Ki-1, NBLST3, Tcrz
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ALK/CD246
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.