Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human alpha 1-Acid Glycoprotein His Protein
SDP

Catalog No. NB172120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172120 50 μg
NB172119 500 μg
NB172118 250 μg
NB172117 20 μg
NB172116 10 μg
NB172115 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. NB172120 Supplier Novus Biologicals™ Supplier No. NBP31835850UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Concentration 1 mg/ml
For Use With (Application) SDS-PAGE
Formulation Phosphate-Buffered Saline (pH 7.4), 10% glycerol
Gene ID (Entrez) 5004
Molecular Weight (g/mol) M.W. Theoretical: 22.4 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias AGP 1, AGP1alpha-1-acid glycoprotein 1, AGP-A, OMD 1, ORM, orosomucoid 1, orosomucoid-1
Gene Symbol ORM1
Research Category Cancer, Immunology, Innate Immunity
Purity or Quality Grade >90%, by SDS-PAGE
Protein alpha 1-Acid Glycoprotein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.