Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human AMCase/CHIA Protein

Catalog No. 9859GH050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-59G-H050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-59G-H050 Supplier R&D Systems™ Supplier No. 9859GH050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave 4-Methylumbelliferyl-beta-D-N,N’,N"e;-triacetylchitotriose (4-MU-TACT). The specific activity is >125 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q9BZP6-1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 27159
Molecular Weight (g/mol) M.W. (Observed): 39-47 kDa, reducing conditions; M.W. (theoretical): 44 kDa
Name acidic mammalian chitinase, AMCASE, CHIA, CHIT2, CHIT2DKFZp313J1722, chitinase, acidic, EC 3.2.1.14, Lung-Specific Protein TSA1902, TSA1902AMCase
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human AMCase/CHIA protein Tyr22-Pro408, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein AMCase/CHIA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.