Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human AMIGO Protein
SDP

Catalog No. 2565AM050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2565AM050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2565AM050 Supplier R&D Systems™ Supplier No. 2565AM050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to enhance survival of E16-E18 rat embryonic cortical neurons.

The ED50 for this effect is 0.6-2.4 μg/mL.

Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Optimal neurite outgrowth is observed between 1.25-10 μg/mL.

Specifications

Accession Number Q86WK6
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 57463
Molecular Weight (g/mol) M.W. (Observed): 50-65 kDa, reducing conditions; M.W. (theoretical): 39.5 kDa
Name AMIGO
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human AMIGO protein Gly28-His367, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol AMIGO1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein AMIGO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.