Learn More
R&DSystems™ Recombinant Human Aminopeptidase N/CD13 His Avi Protein, CF
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 290 |
| Molecular Weight (g/mol) | M.W.-observed: 116-130 kDa, under reducing conditions., M.W.-theroretical: 105 kD |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Aminopeptidase N/CD13 protein Human APN (Lys69-Lys967) (N-terminus) Accession # AAA51719.1 6-His tag Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, ANPEP, AP-M, APN, AP-N, CD13, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPN, PEPNhAPN |
| Conjugate | Biotin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.