Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Angiopoietin-2 Protein
SDP

Catalog No. 623AN025CF Shop All Novus Biologicals Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl, CHAPS and PEG with Trehalose. 1 MG pack size is supplied as a 0.2 μm filtered solution in MOPS, NaCl, CHAPS and PEG.
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, CHAPS and PEG with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
623AN025CF Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl, CHAPS and PEG with Trehalose. 1 MG pack size is supplied as a 0.2 μm filtered solution in MOPS, NaCl, CHAPS and PEG.
623AN025 Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, CHAPS and PEG with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 623AN025CF Supplier R&D Systems™ Supplier No. 623AN025/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Angiopoietin-2 Protein is derived from NS0. The Recombinant Human Angiopoietin-2 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl, CHAPS and PEG with Trehalose. 1 MG pack size is supplied as a 0.2 μm filtered solution in MOPS, NaCl, CHAPS and PEG.
Gene ID (Entrez) 285
Molecular Weight (g/mol) 50.1 kDa
Name Angiopoietin-2
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived human Angiopoietin-2 protein Tyr19-Phe496,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 3 months, -20 to -70°C under sterile conditions after reconstitution. 1 month, 2 to 8°C under sterile conditions after reconstitution.
Structural Form Oligomer
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.