Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Angiopoietin-like 4 Biotin Protein

Catalog No. BT4487050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT4487050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT4487050 Supplier R&D Systems™ Supplier No. BT4487050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Angiopoietin-like 4 Biotin Protein is derived from NS0. The Recombinant Human Angiopoietin-like 4 Biotin Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9BY76
For Use With (Application) Bioactivity
Gene ID (Entrez) 51129
Molecular Weight (g/mol) Theoretical M.W.: 44kDa (unlabeled)
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Angiopoietin-like Protein 4/ANGPTL4 protein Gly26-Ser406 (Lys163Ala,Arg164Ala),with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein
Common Name Angiopoietin-like Protein 4/ANGPTL4
Gene Symbol ANGPTL4
Biological Activity Measured by its ability to promote the expansion of E16 rat liver mononuclear cells in vitro, in the presence of Recombinant Mouse SCF/c-kit Ligand, Recombinant Mouse Thrombopoietin/Tpo, and Recombinant Mouse Flt-3 Ligand. The ED50 for this effect is typically 100-600ng/mL in the presence of a cross-linking antibody, His Tag MAb, Mouse Anti-polyHistidine Monoclonal Antibody.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Gly26-Ser406 (Lys163Ala, Arg164Ala), with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.