Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human APE T7 Protein
SDP

Catalog No. NBP17228050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17228050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17228050 Supplier Novus Biologicals™ Supplier No. NBP17228050UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal T7-tag and corresponding to the amino acids 1-318 of Human APE The Recombinant Human APE Protein is derived from E. coli. The Recombinant Human APE Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number AAH19291
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH8.0) containing 0.2M NaCl, 2mM DTT, 10% glycerol
Gene ID (Entrez) 328
Molecular Weight (g/mol) 36.9kDa
Name APE Protein
Purification Method SDS-PAGE
Quantity 50 μg
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.