Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human AS3MT Protein

Catalog No. 9827MT050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-27M-T050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-27M-T050 Supplier R&D Systems™ Supplier No. 9827MT050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human AS3MT Protein is derived from E. coli. The Recombinant Human AS3MT Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9HBK9-1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Tris and NaCl with Trehalose.
Gene ID (Entrez) 57412
Molecular Weight (g/mol) 42 kDa
Name arsenic (+3 oxidation state) methyltransferase, Arsenic Methyltransferase, arsenite methyltransferase, CYT19, CYT19methyltransferase cyt19, EC 2.1.1.137, Methylarsonite methyltransferase, Methyltransferase Cyt19, S-Adenosyl-L-Methionine:Arsenic(III) Methyltransfe, S-adenosyl-L-methionine:arsenic(III) methyltransferase, S-Adenosylmethionine:Arsenic (III) Methyltransfera, S-adenosylmethionine:arsenic (III) methyltransferase
Quantity 50 μg
Source E. coli-derived human AS3MT protein Ala2-Cys375,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in water.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.