Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ASGR1/ASGPR1 Fc Chimera Protein

Catalog No. 10255AS050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-255-AS050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-255-AS050 Supplier R&D Systems™ Supplier No. 10255AS050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

  • When Recombinant Human ASGR1/ASGPR1 Fc Chimera is immobilized at 2.5 μg/mL (100 μL/well) on a Goat Anti-Human IgG Fc antibody coated plate, the concentration of biotinylated beta-Gal-NAc-PAA that produces 50% of the optimal binding response is 0.75-6 μg/mL.
  • Measured by its binding ability in a functional ELISA.
  • Immobilized Recombinant Human ASGR1/ASGPR1 Fc Chimera binds human plasma von Willebrand Factor with an ED50 of 0.15 to 0.9 μg/mL.

Specifications

Accession Number P07306-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 432
Molecular Weight (g/mol) 59 to 71 kDa, under reducing conditions
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human ASGR1/ASGPR1 protein. MDHuman IgG1 (Pro100-Lys330)IEGRHuman ASGR1 (Asn63-Leu291) Accession # P07306-1N-terminus  C-terminus
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Purity or Quality Grade > 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.