Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ASIP/Agouti Protein

Catalog No. 9094AG050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9094AG050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9094AG050 Supplier R&D Systems™ Supplier No. 9094AG050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human ASIP/Agouti Protein is derived from HEK293. The Recombinant Human ASIP/Agouti Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P42127
For Use With (Application) Bioactivity
Gene ID (Entrez) 434
Molecular Weight (g/mol) Observed M.W.: 18-23kDa, reducing conditions, Theoretical M.W.: 12kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human ASIP protein His23-Cys132
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias agouti (mouse)-signaling protein, agouti signaling protein, agouti signaling protein, nonagouti homolog, agouti signaling protein, nonagouti homolog (mouse), Agouti switch protein, agouti-signaling protein, AGSW, AGTI, AGTIL, AGTILMGC126092, ASPAGSW, MGC126093, nonagouti homolog, SHEP9
Common Name ASIP
Gene Symbol ASIP
Biological Activity Measured by its ability to inhibit alpha-MSH induced eumelanin production in B16F1 mouse melanoma cells. The ED50 for this effect is typically 0.8-4μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived His23-Cys132
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.