Learn More
R&D Systems™ Recombinant Human B7-1/CD80 His-tag Alexa Fluor™ 488 Protein
Description
Specifications
Specifications
| Accession Number | P33681.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 941 |
| Molecular Weight (g/mol) | M.W.-observed: 45-58 kDa, under reducing conditions., M.W.-theroretical: 25 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human B7-1/CD80 protein Val35-Asn242, with a C-terminal 6-His tag |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.