Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human B7-2/CD86 Fc Chimera (CHO-expressed)

Catalog No. 7625B201M
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-25B-201M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 76-25B-201M Supplier R&D Systems™ Supplier No. 7625B201M
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New rh B7-2 protein is Available! It is HEK293 expressed with a His-tag!

Measured by its ability to induce IL-2 secretion by Jurkat human acute T cell leukemia cells. Freeman, G.J. et al. (1993) Science 262:909. The ED50 for this effect is 0.04-0.2 μg/mL.

Specifications

Accession Number AAB03814.1
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 942
Molecular Weight (g/mol) M.W. (Observed): 75-100 kDa, reducing conditions; M.W. (theoretical): 52 kDa (monomer)
Quantity 1 mg
Source Chinese Hamster Ovary cell line, CHO-derived human B7-2/CD86 protein Human B7-2/CD86 (Leu20-His239) (N-terminus) Accession # AAB03814.1 DIEGRMD Human IgG1 (Pro100-Lys330)N-terminus C-terminμs
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72,mgC34413, T-lymphocyte activation antigen CD86
Gene Symbol CD86
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-2/CD86
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.