Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human B7-2/CD86 Fc Chimera Protein

Catalog No. 141B2100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
141B2100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 141B2100 Supplier R&D Systems™ Supplier No. 141B2100
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New rh B7-2 is Available! It has higher purity, ∼3 fold better activity, and is HEK293 expressed!

Measured by its ability to induce IL-2 secretion by Jurkat human acute T cell leukemia cells. Freeman, G.J. et al. (1993) Science 262:909. The ED50 for this effect is 0.2-0.8 μg/mL in the presence of PHA.

Specifications

Accession Number AAB03814
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 942
Molecular Weight (g/mol) M.W. (Observed): 85-95 kDa, reducing conditions; M.W. (theoretical): 52.7 kDa (monomer)
Name B7-2/CD86 Fc Chimera
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived human B7-2/CD86 protein Human B7-2 (Leu20-His239) (N-terminus) Accession # AAB03814 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD86
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein B7-2/CD86
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.