Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human B7-2/CD86 His Protein
SDP

Catalog No. NB002924 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002924 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002924 Supplier Novus Biologicals™ Supplier No. NBP2774860.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a His-Tag and corresponding to the amino acids 31-247aa of Human B7-2/CD86.

Source: Baculovirus

Amino Acid Sequence:

YFNETADLPC QFANSQNQSL SELVVFWQDQ ENLVLNEVYL GKEKFDSVHS KYMGRTSFDS DSWTLRLHNL QIKDKGLYQC IIHHKKPTGM IRIHQMNSEL SVLANFSQPE IVPISNITEN VYINLTCSSI HGYPEPKKMS VLLRTKNSTI EYDGIMQKSQ DNVTELYDVS ISLSVSFPDV TSNMTIFCIL ETDKTRLLSS PFSIELEDPQ PPPDHIPHHH HHH>

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 942
Molecular Weight (g/mol) 25.6 kDa
Quantity 0.5 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Gene Symbol CD86
Buffer PBS (pH 7.4), 10% glycerol.
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity or Quality Grade >90% SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.