Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human B7-H2 Fc Chimera Alexa Fluor™ 647 Protein

Catalog No. AFR165020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFR165020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFR165020 Supplier R&D Systems™ Supplier No. AFR165020
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by flow cytometry for its ability to bind anti-human B7-H2 Antibody conjugated beads. The concentration of Recombinant Human B7-H2 Fc Chimera Alexa Fluor™ 647 (Catalog # AFR165) that produces 50% of the binding response is 2.50-25.0 ng/mL.

Specifications

Accession Number O75144.2
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 23308
Molecular Weight (g/mol) M.W.-observed: 80-90 kDa, under reducing conditions., M.W.-theroretical: 53.4 kDa (monomer)
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human B7-H2 protein Human B7-H2 (Asp19-Ser258) (C-terminus) Accession # O75144.2 DIEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1 LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Conjugate Alexa Fluor 647
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H2/ICOSLG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.